Order Entry
Belgium
Orders LinkContactUsLinkComponent
Separating or resolving buffer (4X)

Separating or resolving buffer (4X)

Separating or Resolving Buffer, 4X, 500 mL
Catalog # J60181.AP
Separating or resolving buffer (4X)
Catalog # J60181.AP
CAS Number:  
Separating or resolving buffer (4X)
Separating or resolving buffer (4X)

Separating or Resolving Buffer, 4X, 500 mL

CAS Number:  
Price
71,10 €
Log in for availability
1 * 500 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Clear

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    Separating or Resolving Buffer, 4X
  • Cat. No.:
    J60181.AP
  • Physical form:
    Liquid
  • Appearance:
    Clear

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.