Buffer, potassium phosphate, 0,2 M solution
Potassium phosphate, 0,2M buffer solution, pH 8,0, Flacon Verre, 1 L
Supplier: Thermo Fisher Scientific
Buffer, potassium phosphate, 0,2 M solution
Catalog # J60851.K2
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401
Documents
Specifications
Specifications
- Type de conditionnement:Flacon Verre
- Cdt:1 L
- Désignation:Potassium phosphate, 0,2M buffer solution, pH 8,0
- Cat. No.:J60851.K2
- N ° CAS.:7778-77-0
- Physical form:Liquid
- MDL No.:MFCD00011401



