Order Entry
Belgium
Orders LinkContactUsLinkComponent
SSC buffer (20X)

SSC buffer (20X)

SSC buffer, 20X, 2 L
Catalog # J60839.K3
SSC buffer (20X)
Catalog # J60839.K3
CAS Number:  
SSC buffer (20X)
SSC buffer (20X)

SSC buffer, 20X, 2 L

CAS Number:  
Price
85,80 €
Log in for availability
1 * 2 L
Product Details & Documents

Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00132371

Specifications

Specifications

  • Pk:
    2 L
  • Description:
    SSC buffer, 20X
  • Cat. No.:
    J60839.K3
  • Physical form:
    Colourless liquid
  • MDL No.:
    MFCD00132371

Specifications