Order Entry
Belgium
Orders LinkContactUsLinkComponent
SSC buffer (20X)

SSC buffer (20X)

SSC buffer, 20X, 1 L
Catalog # J60839.K2
SSC buffer (20X)
Catalog # J60839.K2
CAS Number:  
SSC buffer (20X)
SSC buffer (20X)

SSC buffer, 20X, 1 L

CAS Number:  
Price
55,70 €
Log in for availability
1 * 1 L
Product Details & Documents

Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00132371

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    SSC buffer, 20X
  • Cat. No.:
    J60839.K2
  • Physical form:
    Colourless liquid
  • MDL No.:
    MFCD00132371

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.