Order Entry
Belgium
Orders LinkContactUsLinkComponent
Laemmli SDS sample buffer, reducing (4X)

Laemmli SDS sample buffer, reducing (4X)

Laemmli SDS Sample Buffer, reducing, 4X, 50 mL
Catalog # J60015.AD
Laemmli SDS sample buffer, reducing (4X)
Catalog # J60015.AD
CAS Number:  
Laemmli SDS sample buffer, reducing (4X)
Laemmli SDS sample buffer, reducing (4X)

Laemmli SDS Sample Buffer, reducing, 4X, 50 mL

CAS Number:  
Price
140,00 €
Log in for availability
1 * 50 mL
Product Details & Documents

Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system

Specifications

Specifications

  • Pk:
    50 mL
  • Description:
    Laemmli SDS Sample Buffer, reducing, 4X
  • Cat. No.:
    J60015.AD
  • Physical form:
    Liquid
  • Appearance:
    Blue

Specifications