Order Entry
Belgium
Orders LinkContactUsLinkComponent
Tween 20 blocking buffer, 1% in PBS (10X)

Tween 20 blocking buffer, 1% in PBS (10X)

Tween 20 Blocking Buffer, 1% in PBS (10X), 2 L
Catalog # J61544.K3
Tween 20 blocking buffer, 1% in PBS (10X)
Catalog # J61544.K3
CAS Number:  
Tween 20 blocking buffer, 1% in PBS (10X)
Tween 20 blocking buffer, 1% in PBS (10X)

Tween 20 Blocking Buffer, 1% in PBS (10X), 2 L

CAS Number:  
Price
194,00 €
Log in for availability
1 * 2 L
Product Details & Documents

Storage Temperature: 2 to 8 °C
Physical Form: Viscous liquid
Appearance: Clear

Specifications

Specifications

  • Pk:
    2 L
  • Description:
    Tween 20 Blocking Buffer, 1% in PBS (10X)
  • Cat. No.:
    J61544.K3
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Viscous liquid
  • Appearance:
    Clear

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.