NP-40 lysis buffer
NP-40 lysis buffer, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Documents
Specifications
Specifications
- Pk:250 mL
- Description:NP-40 lysis buffer
- Cat. No.:J60766.AK
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- Appearance:Colourless
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



