Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, ACES, 0,5 M solution

Buffer, ACES, 0,5 M solution

ACES, 0,5M buffer solution, pH 7,0, Glass bottle, 100 mL
Catalog # J60755.AE
Buffer, ACES, 0,5 M solution
Catalog # J60755.AE
CAS Number:  
Buffer, ACES, 0,5 M solution
Buffer, ACES, 0,5 M solution

ACES, 0,5M buffer solution, pH 7,0, Glass bottle, 100 mL

CAS Number:  
Price
64,10 €
Log in for availability
1 * 100 mL
Product Details & Documents

CAS No.: 7365-82-4
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00008030

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Description:
    ACES, 0,5M buffer solution, pH 7,0
  • Cat. No.:
    J60755.AE
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00008030
  • Appearance:
    Colourless
  • CAS No.:
    7365-82-4

Specifications