Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

NP-40 lysis buffer

NP-40 lysis buffer, 500 mL
Catalog # J60766.AP
NP-40 lysis buffer
Catalog # J60766.AP
CAS Number:  
NP-40 lysis buffer
NP-40 lysis buffer

NP-40 lysis buffer, 500 mL

CAS Number:  
Price
115,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless

Specifications

Specifications

  • Pk:
    500 mL
  • Description:
    NP-40 lysis buffer
  • Cat. No.:
    J60766.AP
  • Storage temperature:
    2°C to 8°C
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications