Order Entry
Belgium
Orders LinkContactUsLinkComponent
MNE buffer, pH 6,5

MNE buffer, pH 6,5

MNE buffer, pH 6.5, 250 mL
Catalog # J60690.AK
MNE buffer, pH 6,5
Catalog # J60690.AK
CAS Number:  
MNE buffer, pH 6,5
MNE buffer, pH 6,5

MNE buffer, pH 6.5, 250 mL

CAS Number:  
Price
34,40 €
Log in for availability
1 * 250 mL
Product Details & Documents

Physical Form: Liquid

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    MNE buffer, pH 6.5
  • Cat. No.:
    J60690.AK
  • Physical form:
    Liquid

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.