Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer, potassium phosphate, 0,2 M solution

Buffer, potassium phosphate, 0,2 M solution

Potassium phosphate, 0,2M buffer solution, pH 7,5, Glass bottle, 500 mL
Catalog # J62239.AP
Buffer, potassium phosphate, 0,2 M solution
Catalog # J62239.AP
CAS Number:  
Buffer, potassium phosphate, 0,2 M solution
Buffer, potassium phosphate, 0,2 M solution

Potassium phosphate, 0,2M buffer solution, pH 7,5, Glass bottle, 500 mL

CAS Number:  
Price
88,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    Potassium phosphate, 0,2M buffer solution, pH 7,5
  • Cat. No.:
    J62239.AP
  • Physical form:
    Liquid
  • MDL No.:
    MFCD00011401
  • CAS No.:
    7778-77-0

Specifications