Order Entry
Belgium
Orders LinkContactUsLinkComponent
Inoculating loops, Microstreaker

Inoculating loops, Microstreaker

Small, 2,26 mm ID, blue handle, 2 µl
Catalog # 391-0472
Inoculating loops, Microstreaker
Catalog # 391-0472
Inoculating loops, Microstreaker
Inoculating loops, Microstreaker

Small, 2,26 mm ID, blue handle, 2 µl

Price
99,10 €
Log in for availability
1 * 5 ST
Product Details & Documents

Inoculation loop of nickel-chromium wire of not more than 60 mm length, conforming to Howie recommendations. Colour coded handle.

Specifications
  • Capacity:
    2 µl
  • Description:
    Small, 2,26 mm ID, blue handle
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.