ContactUsLinkComponent
Counting chamber, neubauer-improved (petroff type), depth 0.01mm, double net ruling, dark line, with clamps, with two cover glasses of 0,4mm tickeness. 1 * 10 items
Catalog # MARI0642011
Supplier:  MARIENFELD
Counting chamber, neubauer-improved (petroff type), depth 0.01mm, double net ruling, dark line, with clamps, with two cover glasses of 0,4mm tickeness. 1 * 10 items
Catalog # MARI0642011
Supplier:  MARIENFELD
Frequently Bought Together