ContactUsLinkComponent
PTR-25 MINI-TUBE ROTATOR WITH TIMER, STANDARD PLATFORM FOR 22 X TUBES UP TO 15MM , 5-30RPM 360 ROTATION, HOLDS UP TO 50ML TUBES 1 * 1 items
Catalog # 444-0777
Supplier:  GRANT INSTRUMENTS
undefined
PTR-25 MINI-TUBE ROTATOR WITH TIMER, STANDARD PLATFORM FOR 22 X TUBES UP TO 15MM , 5-30RPM 360 ROTATION, HOLDS UP TO 50ML TUBES 1 * 1 items
Catalog # 444-0777
Supplier:  GRANT INSTRUMENTS
Frequently Bought Together