ES-80 ORBITAL SHAKER INCUBATOR 250RPM, 20MM ORBIT, TO 80C, 8KG LOAD, UP TO 6 X 1000ML FLASKS 1 * 1 items
444-0780
ES-80 ORBITAL SHAKER INCUBATOR 250RPM, 20MM ORBIT, TO 80C, 8KG LOAD, UP TO 6 X 1000ML FLASKS 1 * 1 items
444-0780
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection