DB-4S DRY BLOCK TERMOSTAT FOR 4 X 8 OR 32 0.2ML MICROTUBES/STRIPS 25-100C, TIMER 1M TO 96 H 1 * 1 items
Catalog # 460-0193
Supplier: GRANT INSTRUMENTS
DB-4S DRY BLOCK TERMOSTAT FOR 4 X 8 OR 32 0.2ML MICROTUBES/STRIPS 25-100C, TIMER 1M TO 96 H 1 * 1 items
Catalog # 460-0193
Supplier: GRANT INSTRUMENTS
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection