Orders LinkContactUsLinkComponent
Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
250 ml, 24/29 NS
Catalog # NEUB0752-15-0250
Flasks, flat bottom with standard ground joint
Catalog # NEUB0752-15-0250
Flasks, flat bottom with standard ground joint
250 ml, 24/29 NS
Price
65,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3,3, amber.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Amber stained glass provides protection for materials sensitive to light.
Specifications
  • Socket size:
    24/29 NS
  • Capacity:
    250 ml
  • Seq:
    03