Orders LinkContactUsLinkComponent
Pipette tips
With collar, 1 - 200 µl, -, Transparent, Pipetman, Eppendorf, Biohit, Socorex, Corning, Oxford, Rainin, Bulk (10×1000)
Catalog # DELT200079
Supplier:   DELTALAB
Pipette tips
Catalog # DELT200079
Supplier:  DELTALAB
Pipette tips
With collar, 1 - 200 µl, -, Transparent, Pipetman, Eppendorf, Biohit, Socorex, Corning, Oxford, Rainin, Bulk (10×1000)
Price
790,00 €
Log in for availability
1 * 10.000 ST
Specifications
  • Packed:
    Bulk (10×1000)
  • Sterile:
    -
  • Volume:
    1 - 200 µl
  • Seq:
    05
  • Tip style:
    With collar
  • Tip colour:
    Transparent
  • For use with:
    Pipetman, Eppendorf, Biohit, Socorex, Corning, Oxford, Rainin