Orders LinkContactUsLinkComponent
Pipette tips
Eppendorf type, graduated, 10 - 300 µl, -, Transparent, Gilson, Brand, Labsystems, Biohit, 10 racks, 96 each
Catalog # DELT200077R
Supplier:   DELTALAB
Pipette tips
Catalog # DELT200077R
Supplier:  DELTALAB
Pipette tips
Eppendorf type, graduated, 10 - 300 µl, -, Transparent, Gilson, Brand, Labsystems, Biohit, 10 racks, 96 each
Price
168,00 €
Log in for availability
1 * 10 ST
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    -
  • Volume:
    10 - 300 µl
  • Seq:
    12
  • Tip style:
    Eppendorf type, graduated
  • Tip colour:
    Transparent
  • For use with:
    Gilson, Brand, Labsystems, Biohit