Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Filtration funnels

30 ml, 144 mm, 82 mm, 3, 8 mm, 80 mm
Catalog # BIBB3750/03
Supplier:   SciLabware
Filtration funnels
Catalog # BIBB3750/03
Supplier:  SciLabware
Filtration funnels
Filtration funnels

30 ml, 144 mm, 82 mm, 3, 8 mm, 80 mm

Price
62,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Pyrex® borosilicate glass.

  • Sintered glass disc
  • Filter paper is not required for filtration of solutions
  • Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
Specifications
  • Porosity:
    3
  • Capacity:
    30 ml
  • Length:
    144 mm
  • Seq:
    05
  • Int.Ø top:
    80 mm
  • Stem O-Ø:
    8 mm
  • Stem length:
    82 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.