ContactUsLinkComponent
HEATING TAPE WBS00202 (100 W/m) 200 W, 1,8 m, max. 200°C Heating tape, 4 heating conductors, silicone insulated, outer braiding: tinned copper wire. Operation only with temperature control. Operating currency 230 VAC power 100 W Max. permissive op 1 * 1 items
Catalog # WINKWBS00202800200
Fournisseur:  WINKLER
HEATING TAPE WBS00202 (100 W/m) 200 W, 1,8 m, max. 200°C Heating tape, 4 heating conductors, silicone insulated, outer braiding: tinned copper wire. Operation only with temperature control. Operating currency 230 VAC power 100 W Max. permissive op 1 * 1 items
Catalog # WINKWBS00202800200
Fournisseur:  WINKLER
Frequently Bought Together