Stoppers, self realasing, PTFE
24/40, 65 mm
Catalog # COWI010.1124
Supplier: COWIE
Jump to:
Product Details & Documents
Fit standard ground glass joints.
With sealing rings and self releasing ring.
Specifications
- NS:24/40
- Height:65 mm