Orders LinkContactUsLinkComponent
White light viewer, LabLite illuminator, ViewOne™
White light viewer, LabLite illuminator, ViewOne™
ViewOne™ LabLite illuminator, for 96-well ELISA plate, Petri dish, SDS-PAGE gel, 384-well plates, VL-1000, 115×170×5 mm
Catalog # EMBIVL-1000
Supplier:   EMBITEC
White light viewer, LabLite illuminator, ViewOne™
Catalog # EMBIVL-1000
Supplier:  EMBITEC
White light viewer, LabLite illuminator, ViewOne™
White light viewer, LabLite illuminator, ViewOne™
ViewOne™ LabLite illuminator, for 96-well ELISA plate, Petri dish, SDS-PAGE gel, 384-well plates, VL-1000, 115×170×5 mm
Price
212,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Compact benchtop illuminator helps to visually identify a specific sample well, complete lanes or rows, to aid in loading 96-well plates.
  • Uniform background for enhanced visualization of colorimetric samples
  • Waterproof top allows for visualizing wet samples such as SDS-PAGE gels or dissection samples
  • Portable and space saving
The ViewOne™ LabLite is a stand-alone bench top solution for visualizing colorimetric sample against a uniform white light background. Labs have used the ViewOne with ELISA plates, colony picking and bacterial plates, hemagglutiation assays and cell culture plates. The waterproof top provides a protected surface for usage with set samples such as Coomassie stained SDS-PAGE gels, zymogram gels, and samples for dissection.
The ViewOne™ LabLite Set includes a tilt stand that offers relief from loading over a plate, and adjusts to 10°, 30°, 45°, making it a practical unit that can also be used in a biosafety cabinet.
Delivery information: The ViewOne™ LabLite set includes VL-1007 Universal tilt stand for viewing angle adjustment. Stand-alone model does not include tilt stand.
Specifications
  • Model:
    VL-1000
  • Description:
    ViewOne™ LabLite illuminator, for 96-well ELISA plate, Petri dish, SDS-PAGE gel, 384-well plates
  • Dimensions:
    115×170×5 mm
  • Seq:
    01