Order Entry
Belgium
Orders LinkContactUsLinkComponent
Buffer tablets, Lovibond®

Buffer tablets, Lovibond®

7, 250 Tab.
Catalog # LOVI515611BT
Supplier:   Lovibond
Buffer tablets, Lovibond®
Catalog # LOVI515611BT
Supplier:  Lovibond
CAS Number:  
Buffer tablets, Lovibond®

7, 250 Tab.

CAS Number:  
Price
56,80 €
Log in for availability
1 * 250 Tablet
Product Details & Documents

Ideal for preparing buffer solutions of 4, 7 and 10 pH units.

pH tablet can be added to 100 ml of distilled or deionised water for preparing solution of specified concentration.

Specifications

Specifications

  • Pk:
    250 Tab.
  • pH value:
    7

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.