Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer tablets, Lovibond®

10, 100 Tab.
  LOVI515600BT
 :   Lovibond
Buffer tablets, Lovibond®
  LOVI515600BT
 :  Lovibond
 :  
Buffer tablets, Lovibond®

10, 100 Tab.

 :  
 
27,70 €
   
1 * 1 EA
 

Ideal for preparing buffer solutions of 4, 7 and 10 pH units.

pH tablet can be added to 100 ml of distilled or deionised water for preparing solution of specified concentration.

 

 

  • Pk:
    100 Tab.
  • pH value:
    10