Orders LinkContactUsLinkComponent
Hardness tablets
Hardness tablets
Hardness Yes/No tablets, AF423 Minikit, 250 Tab.
Catalog # LOVI515361BT
Supplier:   Lovibond
Hardness tablets
Catalog # LOVI515361BT
Supplier:  Lovibond
CAS Number:  
Hardness tablets
Hardness Yes/No tablets, AF423 Minikit, 250 Tab.
CAS Number:  
Price
87,70 €
Log in for availability
1 * 250 Tablet
Product Details & Documents
The speed test is based on reverse titration. After adding a reagent tablet to a calibrated test tube, the water sample is added slowly until the colour of the solution changes (e.g. from red to blue). The user can then obtain the result from the liquid level.
A Yes/No test tells the user whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
A Yes/No test tells the user whether a specific ingredient is present in the water and/or if its concentration is higher or lower than a defined level.
Documents
Specifications
Specifications
  • Pk:
    250 Tab.
  • Description:
    Hardness Yes/No tablets
  • For unit:
    AF423 Minikit