Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Buffer tablets, Lovibond®

4, 100 Tab.
  LOVI515620BT
 :   Lovibond
Buffer tablets, Lovibond®
  LOVI515620BT
 :  Lovibond
 :  
Buffer tablets, Lovibond®

4, 100 Tab.

 :  
 
27,70 €
   
1 * 1 EA
 

Ideal for preparing buffer solutions of 4, 7 and 10 pH units.

pH tablet can be added to 100 ml of distilled or deionised water for preparing solution of specified concentration.

 

 

  • Pk:
    100 Tab.
  • pH value:
    4

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.