ContactUsLinkComponent
PS-M3D, 3D multi-function rocker/rotator 7 tilt, 22mm orbit 1 * 1 items
Catalog # 444-0774
Supplier:  GRANT INSTRUMENTS
PS-M3D, 3D multi-function rocker/rotator 7 tilt, 22mm orbit 1 * 1 items
Catalog # 444-0774
Supplier:  GRANT INSTRUMENTS