Galactitol 99+%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:608-66-2
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064288
- Molecular Weight:182.17 g/mol
- Boiling Pt:275…280 °C (1 mmHg)
- Melting Pt:188…189 °C
- Storage Temperature:Ambient
- EINECS:210-165-2
Documents
ACRO11770
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:187 to 191 °C
- Water:≤1.0 % (K.F.)
- Solubility:(3 % in water) Colorless
- HPLC:≥99.0 % (ELSD)
- Heavy metals (as Pb):≤0.003 %
- Insoluble matter:≤0.01 % (in dilute NH4OH)
Specifications
- Pack type:Glass bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



