Funnels, powder
60 mm, 30 mm, 10 mm
Catalog # KAVA632413102060
Supplier: Kavalier
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
These funnels with short stem and bevelled edges are suitable for transferring powdery materials.
Ordering information: Ø = internal stem diameter.
Specifications
- Ø:10 mm
- Funnel O-Ø:60 mm
- Stem length:30 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



