Orders LinkContactUsLinkComponent
Labels, hazard warning
Labels, hazard warning
Hazard labels: danger, —, Polyester, Black on yellow, 100×87 mm, 3 labels/card
Catalog # 116-0754
Supplier:   Brady
Labels, hazard warning
Catalog # 116-0754
Supplier:  Brady
Labels, hazard warning
Labels, hazard warning
Hazard labels: danger, —, Polyester, Black on yellow, 100×87 mm, 3 labels/card
Price
9,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifications
  • Colour:
    Black on yellow
  • Description:
    Hazard labels: danger
  • Dimensions:
    100×87 mm
  • Material:
    Polyester
  • Model:
  • Packed:
    3 labels/card