Scoops
1100 ml, 360 mm, HDPE, White
LAMA65
These scoops are made of high density PE. Ideal for use in food applications.
- For withdrawal of solid and granulates samples
- Rounded bottom
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
- Capacity:1100 ml
- Colour:White
- Length:360 mm
- Material:HDPE
- Seq:04



