ContactUsLinkComponent
GBOX-CHEMI-HR1.4-E Gel Documentation System, 230V, 50Hz, 16bit system, Image resolution 5.78MP, camera resolution 1.4MP. pixel size 6.45 x 6.45 µm, with motor driven zoom f1.2 lens. 12.5 - 75mm with Feedback (+2 and step-up) 7 Position motor driven f 1 * 1 items
Catalog # SYNOGBOXCHEMIHR14E
GBOX-CHEMI-HR1.4-E Gel Documentation System, 230V, 50Hz, 16bit system, Image resolution 5.78MP, camera resolution 1.4MP. pixel size 6.45 x 6.45 µm, with motor driven zoom f1.2 lens. 12.5 - 75mm with Feedback (+2 and step-up) 7 Position motor driven f 1 * 1 items
Catalog # SYNOGBOXCHEMIHR14E
Frequently Bought Together