Orders LinkContactUsLinkComponent
Micro extraction by packed sorbent (MEPS™)
Micro extraction by packed sorbent (MEPS™)
XCHANGE® syringe, 100 µl, MEPS™ BINs
Catalog # SGEA2928500
Micro extraction by packed sorbent (MEPS™)
Catalog # SGEA2928500
Micro extraction by packed sorbent (MEPS™)
Micro extraction by packed sorbent (MEPS™)
XCHANGE® syringe, 100 µl, MEPS™ BINs
Price
131,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Micro Extraction by Packed Sorbent (MEPS™) is a new development in the field of sample preparation by SPE (solid phase extraction). The MEPS™ product consists of two parts, the MEPS™ syringe and the MEPS™ Barrel Insert and Needle assembly (BIN). The MEPS™ approach to sample preparation is suitable for reversed phases, normal phases, mixed mode and ion exchange chemistries. Because MEPS™ BINs can be used for sample volumes as small as 10 µl it makes them particularly well suited to on-line use with LC-MS analysis of volume limited samples.
  • The BIN contains the SPE phase that is built into the syringe needle
  • The BIN is used with a 100 or 250 µl gas tight MEPS™ syringe that allows fluid handling at normal SPE pressures
  • Sample preparation for complex biological samples is readily adapted to MEPS™ and reduces the volume of sample and reagents required for extraction
Base material is silica with mean particle size of 45 µm and pore size of 60 Å.
Specifications
  • For:
    MEPS™ BINs
  • Volume:
    100 µl
  • Description:
    XCHANGE® syringe
  • Seq:
    06