Analytical column chiralpak® zwix(+) 250 x 4 mm 3 µm 1 * 1 items
Catalog # DAIC51515
Supplier: CHIRAL TECHNOLOGIES
Analytical column chiralpak® zwix(+) 250 x 4 mm 3 µm 1 * 1 items
Catalog # DAIC51515
Supplier: CHIRAL TECHNOLOGIES
Frequently Bought Together
SafetyWasteCaps
Supplier: S.C.A.T.
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
HPLC columns, LiChrospher®
Supplier: Merck
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
VWR®, TLC Aluminium Plates, Silica, Unmodified Layers
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
Vial racks
Supplier: MACHEREY-NAGEL
VWR®, Screw Neck Vials ND15
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Septa 12 mm
Supplier: VWR Collection
Glass capillaries and graduated micropipettes, disposable
Supplier: CAMAG
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
Calibration standard, single element, silicon for AAS and ICP-OES, pure grade
Supplier: PerkinElmer
Caps for 20 ml glass crimp top headspace vials, level 1 everyday analysis, SureSTART™
Supplier: Thermo Fisher Scientific
Removable needles
Supplier: HAMILTON BONADUZ