Orders LinkContactUsLinkComponent
Sample containers, straight, with screw cap, WHEATON®
Sample containers, straight, with screw cap, WHEATON®
100 ml, Plain label, sterile, Clear, PS/metal
Catalog # 216-1012
Supplier:   DWK Life Sciences
Sample containers, straight, with screw cap, WHEATON®
Catalog # 216-1012
Supplier:  DWK Life Sciences
Sample containers, straight, with screw cap, WHEATON®
Sample containers, straight, with screw cap, WHEATON®
100 ml, Plain label, sterile, Clear, PS/metal
Price
199,00 €
Log in for availability
1 * 200 ST
Product Details & Documents
Polystyrene clear, with metal screw cap.
  • Manufactured from FDA approved virgin polystyrene
  • Supplied sterile and for in vitro use only
  • CE marked in accordance with European Directive 98/79/EC
  • All containers are leak tested to British Standard BS5213:1975(1989)
  • All materials are non cytotoxic
Straight walled vessels ideal for sample collection, storage and transport. Polystyrene offers good thermal and chemical resistance. Supplied with flow seal metal cap.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Type:
    Plain label, sterile
  • Capacity:
    100 ml
  • Colour:
    Clear
  • Material:
    PS/metal