Orders LinkContactUsLinkComponent
Cold traps, long version, type KFL
Cold traps, long version, type KFL
Cold trap, KFL 29-OK-A, with spout, complete with Dewar and plastic ring, with spherical joints S 29 and O-ring seal, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap 250 ml and coolant cap 2000 ml
Catalog # KGWA17381
Supplier:   KGW
Cold traps, long version, type KFL
Catalog # KGWA17381
Supplier:  KGW
Cold traps, long version, type KFL
Cold traps, long version, type KFL
Cold trap, KFL 29-OK-A, with spout, complete with Dewar and plastic ring, with spherical joints S 29 and O-ring seal, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap 250 ml and coolant cap 2000 ml
Price
753,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These cold traps (cold finger condensers) are constructed in the same way as standard cold traps.
  • Available with or without spouts
  • Complete versions include Dewar flask and plastic ring
The condensation area of the cold traps has been extended in order to trap more condensation. In addition to this, the complete cold trap versions also have a larger plastic ring and a bigger Dewar flask.
An upgraded version is a long version cold trap with an outlet spout. These cold traps are constructed in the same way as standard cold traps but have a GL 32 glass thread together with a screw-on cap, which is the actual spout. With the help of this spout the condensate can easily be poured out of the cold trap. Therefore the cold trap is easily to clean.
Certifications: Made from borosilicate glass 3.3, in compliance with DIN/ISO 3585 (DURAN made by Schott).
Specifications
  • Description:
    Cold trap, KFL 29-OK-A, with spout, complete with Dewar and plastic ring, with spherical joints S 29 and O-ring seal, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap 250 ml and coolant cap 2000 ml
  • Seq:
    04
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cold trap, KFL SL 29-GL, spare part, without spout, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, condensate cap. 250 ml
    Cold trap, KFL SL 29-GL, spare part, without spout, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, condensate cap. 250 ml
    KGWA17367
    Log in for availability
    1 * 1 ST - 283,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Cold trap SL 29-O-A, spare part, with spout, with spherical joints S 29 and O-ring seal, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml
    Cold trap SL 29-O-A, spare part, with spout, with spherical joints S 29 and O-ring seal, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml
    KGWA17386
    Log in for availability
    1 * 1 ST - 386,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Cold trap SL 29 GL-A, spare part, with spout, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml
    Cold trap SL 29 GL-A, spare part, with spout, with threaded glass joint GL 18 and PTFE olive, borosilicate glass 3.3, with GL 32 glass thread, screw-on cap, condensate cap. 250 ml
    KGWA17387
    Log in for availability
    1 * 1 ST - 347,00 €