Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bosshead, cross type

Bosshead, cross type

With brass thumb screws, 0 - 16 mm, Tempered cast iron
Catalog # NEUBM020-4850
Bosshead, cross type
Catalog # NEUBM020-4850
Bosshead, cross type

With brass thumb screws, 0 - 16 mm, Tempered cast iron

Price
40,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Bosshead made of tempered cast iron, powder-coated.

  • For rods up to 16 mm Ø
  • With brass thumb or stainless steel safety heart screws
  • Thread: M8

Weight: 180 g

Specifications
  • Spanning width:
    0 - 16 mm
  • Description:
    With brass thumb screws
  • Material:
    Tempered cast iron
  • Seq:
    01
  • Cat. No.:
    NEUBM020-4850
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.