ContactUsLinkComponent
THERMOCYCLER GS1 OHNE BLOCK Temperature range: 4°C-99°C Volume range: 2-150 µ l Block accuracy: +/- 0.4°C (20-99°C) Block homogeneity: +/- 0.4°C Ramp rate: Up to 3°CS- 1 Gradient temp range:30°C - 80°C Max/min gradient span: 30°C / 4°C Lid temperatur 1 * 1 items
Catalog # LBTHGS-00001
Supplier:  LABTECH
THERMOCYCLER GS1 OHNE BLOCK Temperature range: 4°C-99°C Volume range: 2-150 µ l Block accuracy: +/- 0.4°C (20-99°C) Block homogeneity: +/- 0.4°C Ramp rate: Up to 3°CS- 1 Gradient temp range:30°C - 80°C Max/min gradient span: 30°C / 4°C Lid temperatur 1 * 1 items
Catalog # LBTHGS-00001
Supplier:  LABTECH
Frequently Bought Together