VWR®, N-Plan Objectives
IOS N-plan achromatic objective, 1,5 mm
Catalog # 630-1641
Supplier: VWR Collection
Jump to:
Product Details & Documents
These objectives are ideal for use in the education field and for laboratory routine applications.
IOS N-Plan objectives feature an infinity-corrected optical system.
Documents
VWR® for Microscopy
Product Literature
Specifications
- Magnification:20×/0,40 mm
- Description:IOS N-plan achromatic objective
- Seq:203
- Working distance:1,5 mm