Order Entry
Belgium
Orders LinkContactUsLinkComponent
J.T.Baker® BAKER ANALYZED®, Buffer Solutions

J.T.Baker® BAKER ANALYZED®, Buffer Solutions

4,00, ±0,02, Phthalate, 25 °C, Plastic bottle, 2,5 L
Catalog # 5606.2500PE
Supplier:   Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
Catalog # 5606.2500PE
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
J.T.Baker® BAKER ANALYZED®, Buffer Solutions

4,00, ±0,02, Phthalate, 25 °C, Plastic bottle, 2,5 L

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 2,5 L
Product Details & Documents

Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilise quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control.

Contain 0,01% Sodium benzoate.

Certifications: This buffer is traceable to NIST Standard reference buffer.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Phthalate
  • Pk:
    2,5 L
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0,02
  • pH value:
    4,00

Specifications