Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

J.T.Baker® BAKER ANALYZED®, Buffer Solutions

7,00, ±0,02, Phosphate, 20 °C, Plastic container, 10 L
Catalogus # 5608.9010
Leverancier:   Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
Catalogus # 5608.9010
Leverancier:  Avantor
CAS-nummer:  
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
J.T.Baker® BAKER ANALYZED®, Buffer Solutions

7,00, ±0,02, Phosphate, 20 °C, Plastic container, 10 L

CAS-nummer:  
Price
254,13 €
Log in voor beschikbaarheid
1 * 10 L
Product Details & Documents

Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilise quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control.

Contain 0,01% Sodium benzoate.

 : This buffer is traceable to NIST Standard reference buffer.

Specificaties

Specificaties

  • Pack type:
    Plastic container
  • pH buffer type:
    Phosphate
  • Pk:
    10 L
  • Reference temperature:
    20 °C
  • pH accuracy:
    ±0,02
  • pH value:
    7,00

Specificaties