Order Entry
Belgium
Orders LinkContactUsLinkComponent
J.T.Baker® BAKER ANALYZED®, Buffer Solutions

J.T.Baker® BAKER ANALYZED®, Buffer Solutions

3,00, ±0,02, Citrate, 20 °C, Plastic bottle, 1 L
Catalog # 5643.1000
Supplier:   Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
Catalog # 5643.1000
Supplier:  Avantor
CAS Number:  
J.T.Baker® BAKER ANALYZED®, Buffer Solutions
J.T.Baker® BAKER ANALYZED®, Buffer Solutions

3,00, ±0,02, Citrate, 20 °C, Plastic bottle, 1 L

CAS Number:  
Price
73,99 €
Log in for availability
1 * 1 L
Product Details & Documents

Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilise quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardisation of solutions, helping streamline high-volume analytical work in applications such as quality control.

Contain 0,01% Sodium benzoate.

Certifications: This buffer is traceable to NIST Standard reference buffer.

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Citrate
  • Pk:
    1 L
  • Reference temperature:
    20 °C
  • pH accuracy:
    ±0,02
  • pH value:
    3,00

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.