J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
7,00, ±0,02, Phosphate, Green, 20 °C, 500 mL
Fournisseur: Avantor
J.T.Baker® BAKER ANALYZED®, Buffer Solutions, Colour Coded
N° de catalogue 5647.0500
Fournisseur: Avantor
Numéro CAS:
Aller à :
Product Details & Documents
The quality of many BAKER ANALYZED™ reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS).
- Colour coded
- Potassium hydrogen phthalate based pH 4 buffer (red)
- Phosphate based pH 7 buffer (yellow)
- Borate based pH 10 buffer (blue)
Ready-to-use pH buffers are manufactured in large lots to stringent specifications and utilize quality control procedures to reduce lot-to-lot variability. You can eliminate the time and expense of preparation and standardization of solutions, helping streamline high-volume analytical work in applications such as quality control.
Solutions are available in both clear and colour-coded varieties.
A certificate is provided to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Certifications: Each lot compared to NIST Standard Reference Buffer.
Documents
Certificats
Rechercher par numéro de lotChemical Mining Supplies
Documentation produit
spec_BAKR5647.0500.pdf spec_BAKR5647.1000.pdf spec_BAKR5647.9010.pdf spec_BAKR5655-01 spec_BAKR5656-03
Spécification du produit
Plus
Spécifications
Spécifications
- pH buffer type:Phosphate
- Pk:500 mL
- Reference temperature:20 °C
- Colour:Green
- pH accuracy:±0,02
- pH value:7,00



