ContactUsLinkComponent
Chromaster HPLC Semi-Micro Retrofit kit for 600 bar applications includes: slip-free capillary 400 mm HITA891-3805, Flex capillary 280 mm x 0.12 ID HEWL5021-1822, 1x fitting UPCHU-400, 1x ferrule UPCHU-401, 1x Fingertight Fitting UPCHF-127, 1x samp R 1 * 1 items
Catalog # HPLCCM600SMKITUNI
Chromaster HPLC Semi-Micro Retrofit kit for 600 bar applications includes: slip-free capillary 400 mm HITA891-3805, Flex capillary 280 mm x 0.12 ID HEWL5021-1822, 1x fitting UPCHU-400, 1x ferrule UPCHU-401, 1x Fingertight Fitting UPCHF-127, 1x samp R 1 * 1 items
Catalog # HPLCCM600SMKITUNI
Frequently Bought Together