Clarithromycin Impurities, Reference Standards
Clarithromycin Impurity R, USP reference standard, Vial, 10 mg
Catalog # 1134642.
Supplier: USP
Aller à :
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Spécifications
Spécifications
- Pack type:Vial
- Pk:10 mg
- Standards:USP reference standard
- Description:Clarithromycin Impurity R
- Cat. No.:1134642.
- Formula:C₃₆H₆₅NO₁₃
- Description:Clarithromycin Impurity R
- Synonym(s):3"-N-Demethylerythromycin A
- CAS No.:992-62-1



