Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cilostazol, Reference Standards

Cilostazol, Reference Standards

Cilostazol Related Compound B, USP reference standard, Vial, 50 mg
  1134175.
 :   USP
Cilostazol, Reference Standards
  1134175.
 :  USP
 :  
Cilostazol, Reference Standards

Cilostazol Related Compound B, USP reference standard, Vial, 50 mg

 :  
 
1 150,00 €
   
1 * 50 mg
 

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

 

 

  • Pack type:
    Vial
  • Pk:
    50 mg
  • Standards:
    USP reference standard
  • Description:
    Cilostazol Related Compound B
  • Cat. No.:
    1134175.
  • Formula:
    C₂₀H₂₅N₅O₂
  • Description:
    Cilostazol Related Compound B
  • Synonym(s):
    6-[4-(1-Cyclohexyl-1H-tetrazol-5-yl)-butoxy]-1H-quinolin-2-one
  • CAS No.:
    73963-62-9

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.