Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Methotrexate, Reference Standards

Methotrexate System Suitability Mixture, USP reference standard, Vial, 5 mg
Catalogus # 1413999.
Leverancier:   USP
Methotrexate, Reference Standards
Catalogus # 1413999.
Leverancier:  USP
CAS-nummer:  
Methotrexate, Reference Standards

Methotrexate System Suitability Mixture, USP reference standard, Vial, 5 mg

CAS-nummer:  
Price
1 410,00 €
Log in voor beschikbaarheid
1 * 5 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specificaties

Specificaties

  • Pack type:
    Vial
  • Pk:
    5 mg
  • Standards:
    USP reference standard
  • Description:
    Methotrexate System Suitability Mixture
  • UN No.:
    2811
  • Cat. No.:
    1413999.
  • Description:
    Methotrexate

Specificaties