ContactUsLinkComponent
CryoCeps Cryogenic Vial Grippers, green. Tool to access and extract frozen cryogenic vials and microtubes from CoolRack or CoolCell. 1 * 2 items
Catalog # BCISBCS-213
Supplier:  BIOCISION
undefined
CryoCeps Cryogenic Vial Grippers, green. Tool to access and extract frozen cryogenic vials and microtubes from CoolRack or CoolCell. 1 * 2 items
Catalog # BCISBCS-213
Supplier:  BIOCISION
Frequently Bought Together