Labels, hazard warning
Hazard labels: biological hazard, PIC W009-TRI 025-PE-ROLL/1, Polyester, Black on yellow, 25×18 mm, 250 labels/roll
Catalogus # BRDY827646
Leverancier: Brady
Ga naar:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certificeringen: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Documents
Specificaties
- Colour:Black on yellow
- Description:Hazard labels: biological hazard
- Dimensions:25×18 mm
- Material:Polyester
- Seq:05
- Model:PIC W009-TRI 025-PE-ROLL/1
- Packed:250 labels/roll



